4-Amino-3,5-xylenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-3,5-dimethylphenol
OR904670-100G
APOSOR904670-100GEA 365 EUR
APOSOR904670-100G
4-Amino-3,5-xylenol 98%
4-Amino-3,5-xylenol
Formula: C₈H₁₁NO
MW: 137.1800 g/mol
Melting Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD01741459
CAS Number: 3096-70-6