3-Methyl-1H-pyrazolo[4,3-b]pyridine 95%
Supplier: Apollo Scientific
OR904805-1G OR904805-250MG
APOSOR904805-1GEA 806 EUR
APOSOR904805-1G APOSOR904805-250MG
3-Methyl-1H-pyrazolo[4,3-b]pyridine 95%
3-Methyl-1H-pyrazolo[4,3-b]pyridine
Formula: C7H7N3
MW: 133.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062968
CAS Number: 194278-45-0