2-(6-Chloropyrazin-2-yl)acetic acid 95%
Supplier: Apollo Scientific
OR905690-1G OR905690-250MG
APOSOR905690-1GEA 219 EUR
APOSOR905690-1G APOSOR905690-250MG
2-(6-Chloropyrazin-2-yl)acetic acid 95%
2-(6-Chloropyrazin-2-yl)acetic acid
Formula: C₆H₅ClN₂O₂
MW: 172.570 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907855
CAS Number: 930798-25-7