4,7-Dimethoxy-1,10-phenanthroline 97%
Supplier: Apollo Scientific
OR905864-5G
APOSOR905864-5GEA 68.3 EUR
APOSOR905864-5G
4,7-Dimethoxy-1,10-phenanthroline 97%
4,7-Dimethoxy-1,10-phenanthroline
Formula: C₁₄H₁₂N₂O₂
MW: 240.2573 g/mol
Melting Pt: 197…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233883
CAS Number: 92149-07-0