2-Chloro-4-methoxypyrimidine-5-carbonitrile 98%
Supplier: Apollo Scientific
OR905966-1G OR905966-250MG
APOSOR905966-1GEA 179 EUR
APOSOR905966-1G APOSOR905966-250MG
2-Chloro-4-methoxypyrimidine-5-carbonitrile 98%
2-Chloro-4-methoxypyrimidine-5-carbonitrile
Formula: C₆H₄ClN₃O
MW: 169.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250650
CAS Number: 1106295-93-5