1,2-Benzisothiazol-3(2H)-one 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,2-Benzisothiazol-3-one
OR906447-100G OR906447-25G
APOSOR906447-100GEA 94.4 EUR
APOSOR906447-100G APOSOR906447-25G
1,2-Benzisothiazol-3(2H)-one 98%
1,2-Benzisothiazol-3(2H)-one
Formula: C₇H₅NOS
MW: 151.19 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044001
CAS Number: 2634-33-5
UN: 3077
ADR: 9+EHS III