4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 98%
Supplier: Apollo Scientific
Warning
Synonyms: (Chloro(4-methoxyphenyl)methylene)dibenzene, Mmt-Cl
OR906880-100G OR906880-500G
APOSOR906880-100GEA 68.3 EUR
APOSOR906880-100G APOSOR906880-500G
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 98%
4-Methoxytrityl chloride (p-Anisylchlorodiphenylmethane)
Formula: C₂₀H₁₇ClO
MW: 308.8 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000814
CAS Number: 14470-28-1