Methyl-3-ethyl-1H-pyrazole-5-carboxylate 95%
Supplier: Apollo Scientific
Synonyms: Methyl 3-ethyl-1H-pyrazole-5-carboxylate, 3-Ethyl-5-pyrazolcarboxylic acid methyl ester
OR906998-10G OR906998-1G OR906998-5G
APOSOR906998-10GEA 337 EUR
APOSOR906998-10G APOSOR906998-1G APOSOR906998-5G
Methyl-3-ethyl-1H-pyrazole-5-carboxylate 95%
Methyl-3-ethyl-1H-pyrazole-5-carboxylate
Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236689
CAS Number: 834869-10-2