(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 95%
Supplier: Apollo Scientific
Synonyms: (S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, (2S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
OR907114-1G OR907114-250MG
APOSOR907114-1GEA 133 EUR
APOSOR907114-1G APOSOR907114-250MG
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 95%
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
Formula: C₉H₁₆N₂O₂
MW: 184.2355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066229
CAS Number: 78342-42-4