2-Cyano-4-methoxybenzaldehyde 95%
Supplier: Apollo Scientific
OR907192-1G OR907192-5G
APOSOR907192-1GEA 94.4 EUR
APOSOR907192-1G APOSOR907192-5G
2-Cyano-4-methoxybenzaldehyde 95%
2-Cyano-4-methoxybenzaldehyde
Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD08275456
CAS Number: 21962-47-0