4,5-Dichloropyrimidin-2-amine 96%

Supplier: Apollo Scientific

OR907201-1G OR907201-5G
APOSOR907201-1GEA 30.6 EUR
APOSOR907201-1G APOSOR907201-5G
4,5-Dichloropyrimidin-2-amine 96%
4,5-Dichloropyrimidin-2-amine
Formula: C₄H₃Cl₂N₃
MW: 163.9900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193667
CAS Number: 403854-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR