2-(Trimethylsiloxy)-1,3-butadiene 96%
Supplier: Apollo Scientific
Danger
Synonyms: (Buta-1,3-dien-2-yloxy)trimethylsilane
OR907328-25G OR907328-5G
APOSOR907328-25GEA 377 EUR
APOSOR907328-25G APOSOR907328-5G
2-(Trimethylsiloxy)-1,3-butadiene 96%
2-(Trimethylsiloxy)-1,3-butadiene
Formula: C₇H₁₄OSi
MW: 142.27 g/mol
Boiling Pt: 55 °C
Density: 0.82 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009852
CAS Number: 38053-91-7
UN: 1993
ADR: 3,II