Methyl-3-bromo-5-hydroxybenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-bromo-5-hydroxybenzoate
OR907586-5G
APOSOR907586-5GEA 74.1 EUR
APOSOR907586-5G
Methyl-3-bromo-5-hydroxybenzoate 98%
Methyl 3-bromo-5-hydroxybenzoate
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD10566800
CAS Number: 192810-12-1