Methyl-4-(dimethylamino)benzoate 98%
Supplier: Apollo Scientific
Synonyms: 4-(Dimethylamino)benzoic acid methyl ester, Methyl 4-dimethylaminobenzoate
OR907612-100G OR907612-25G OR907612-5G
APOSOR907612-100GEA 266 EUR
APOSOR907612-100G APOSOR907612-25G APOSOR907612-5G
Methyl-4-(dimethylamino)benzoate 98%
Methyl 4-(dimethylamino)benzoate
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014905
CAS Number: 1202-25-1