2-Carboxycinnamic acid 95%

Supplier: Apollo Scientific

APOSOR908071-1GEA 102 EUR
APOSOR908071-1G APOSOR908071-5G
2-Carboxycinnamic acid 95%
2-Carboxycinnamic acid
Formula: C₁₀H₈O₄
MW: 192,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004380
CAS Number: 612-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR