2,6-Diisopropylphenylboronic acid 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Diisopropylphenylboronic Acid
OR908515-1G OR908515-5G
APOSOR908515-1GEA 33.5 EUR
APOSOR908515-1G APOSOR908515-5G
2,6-Diisopropylphenylboronic acid 96%
2,6-Diisopropylphenylboronic acid
Formula: C₁₂H₁₉BO₂
Melting Pt: 167…172 °C
Storage Temperature: Ambient
MDL Number: MFCD16876786
CAS Number: 363166-79-4