Triphenylphosphine 99+%
Supplier: Apollo Scientific
Warning
OR9088-1KG OR9088-250G OR9088-5KG
APOSOR9088-1KGEA 78.3 EUR
APOSOR9088-1KG APOSOR9088-250G APOSOR9088-5KG
Triphenylphosphine 99+%
Triphenylphosphine
Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 360 °C
Melting Pt: 78…82 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873