1-Benzhydrylazetidin-3-one 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Benzhydryl-3-azetidinone, 1-Diphenylmethyl-3-azetidinone
OR9090-5G OR9090-1G OR9090-10G
APOSOR9090-5GEA 27.7 EUR
APOSOR9090-5G APOSOR9090-1G APOSOR9090-10G
1-Benzhydrylazetidin-3-one 98%
1-Benzhydrylazetidin-3-one
Formula: C₁₆H₁₅NO
MW: 237.30 g/mol
Boiling Pt: 350…355 °C
Density: 1.182 g/cm³ (25 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00964539
CAS Number: 40320-60-3