3-(2-Methoxyphenyl)propionic acid 98%
Supplier: Apollo Scientific
Synonyms: 3-(2-Methoxyphenyl)propanoic acid
OR909100-100G OR909100-25G
APOSOR909100-100GEA 256 EUR
APOSOR909100-100G APOSOR909100-25G
3-(2-Methoxyphenyl)propionic acid 98%
3-(2-Methoxyphenyl)propionic acid
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00002772
CAS Number: 6342-77-4