1-(4-Chlorophenyl)cyclopropanecarbonitrile 98%
Supplier: Apollo Scientific
OR909221-100G OR909221-25G
APOSOR909221-100GEA 409 EUR
APOSOR909221-100G APOSOR909221-25G
1-(4-Chlorophenyl)cyclopropanecarbonitrile 98%
1-(4-Chlorophenyl)cyclopropanecarbonitrile
Formula: C₁₀H₈ClN
MW: 177.6302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019211
CAS Number: 64399-27-5