tert-Butyl-3-oxoazetidine-1-carboxylate 98%
Supplier: Apollo Scientific
Danger
Synonyms: N-Boc-3-azetidinone, Tert-butyl 3-oxoazetidine-1-carboxylate, tert-Butyl 3-oxoazetidine-1-carboxylate, 1-BOC-3-Azetidinone
OR9093-25G OR9093-5G OR9093-100G
APOSOR9093-25GEA 40.6 EUR
APOSOR9093-25G APOSOR9093-5G APOSOR9093-100G
tert-Butyl-3-oxoazetidine-1-carboxylate 98%
tert-Butyl-3-oxoazetidine-1-carboxylate
Formula: C₈H₁₃NO₃
MW: 171.2 g/mol
Melting Pt: 47…52 °C
Storage Temperature: Ambient
MDL Number: MFCD01861741
CAS Number: 398489-26-4
UN: 3335
ADR: 9 III