5-Chloro-1,3-dihydro-2H-benzimidazol-2-one 95%
Supplier: Apollo Scientific
Synonyms: 5-Chloro-1H-benzo[d]imidazol-2(3H)-one
OR909307-5G
APOSOR909307-5GEA 91.5 EUR
APOSOR909307-5G
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one 95%
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one
Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00493866
CAS Number: 2034-23-3