1-Isopropyl-1H-pyrazole-4-carbaldehyde 98%
Supplier: Apollo Scientific
OR909353-1G OR909353-5G
APOSOR909353-1GEA 42.2 EUR
APOSOR909353-1G APOSOR909353-5G
1-Isopropyl-1H-pyrazole-4-carbaldehyde 98%
1-Isopropyl-1H-pyrazole-4-carbaldehyde
Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06260724
CAS Number: 313735-67-0