98%

Supplier: Apollo Scientific

Synonyms: N-Phenylpiperidine

OR909425-1G OR909425-25G OR909425-5G
APOSOR909425-1GEA 56.7 EUR
APOSOR909425-1G APOSOR909425-25G APOSOR909425-5G
98%
1-Phenylpiperidine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129777
CAS Number: 4096-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR