Ethyl-5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxylate 98%
Supplier: Apollo Scientific
OR909675-1G OR909675-5G
APOSOR909675-1GEA 219 EUR
APOSOR909675-1G APOSOR909675-5G
Ethyl-5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxylate 98%
Ethyl-5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxylate
Formula: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15072122
CAS Number: 1223748-44-4