2-(4-Bromo-1H-pyrazol-1-yl)ethanol 97%
Supplier: Apollo Scientific
Synonyms: 4-Bromo-1-(2-hydroxyethyl)pyrazole
OR909814-10G OR909814-1G OR909814-5G
APOSOR909814-10GEA 539 EUR
APOSOR909814-10G APOSOR909814-1G APOSOR909814-5G
2-(4-Bromo-1H-pyrazol-1-yl)ethanol 97%
2-(4-Bromo-1H-pyrazol-1-yl)ethanol
Formula: C₅H₇BrN₂O
MW: 191.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06804929
CAS Number: 214614-81-0