5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole 98%

Supplier: Apollo Scientific

Synonyms: 5-Methyl-3-p-tolyl-1,2,4-oxadiazole

OR909824-10G OR909824-1G OR909824-5G
APOSOR909824-10GEA 538 EUR
APOSOR909824-10G APOSOR909824-1G APOSOR909824-5G
5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole 98%
5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₁₀N₂O
MW: 174.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085801
CAS Number: 81386-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR