5-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 98%
Supplier: Apollo Scientific
OR909943-1G OR909943-250MG OR909943-5G
APOSOR909943-1GEA 270 EUR
APOSOR909943-1G APOSOR909943-250MG APOSOR909943-5G
5-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 98%
5-Methylimidazo[1,2-a]pyridine-3-carbaldehyde
Formula: C₉H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD11053823
CAS Number: 178488-37-4