Ethyl 2-oxo-2,3-dihydrooxazole-5-carboxylate 98%
Supplier: Apollo Scientific
OR910103-1G OR910103-5G
APOSOR910103-1GEA 219 EUR
APOSOR910103-1G APOSOR910103-5G
Ethyl 2-oxo-2,3-dihydrooxazole-5-carboxylate 98%
Ethyl 2-oxo-2,3-dihydrooxazole-5-carboxylate
Formula: C₆H₇NO₄
MW: 157.120 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026055
CAS Number: 1150271-25-2