4-Bromo-1,5-dimethyl-1H-pyrazole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1,5-dimethylpyrazole
OR910220-1G OR910220-5G OR910220-10G
APOSOR910220-1GEA 58 EUR
APOSOR910220-1G APOSOR910220-5G APOSOR910220-10G
4-Bromo-1,5-dimethyl-1H-pyrazole 95%
4-Bromo-1,5-dimethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175.0265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759006
CAS Number: 5775-86-0