5-Methoxy-o-toluidine 98%
Supplier: Apollo Scientific
Synonyms: 2-Amino-4-methoxytoluene, 3-Amino-4-methylanisole, 1-Amino-3-methoxy-6-methylbenzene, 6-Methyl-m-anisidine, 5-Methoxy-2-methylaniline
OR910498-100G OR910498-25G
APOSOR910498-100GEA 603 EUR
APOSOR910498-100G APOSOR910498-25G
5-Methoxy-o-toluidine 98%
5-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 253
Melting Pt: 47
Density: 1.39
Storage Temperature: Ambient
MDL Number: MFCD00075057
CAS Number: 50868-72-9