Methyl-1-methyl-1H-1,3-benzimidazole-5-carboxylate 97%
Supplier: Apollo Scientific
Synonyms: Methyl 1-methyl-1H-benzo[d]imidazole-5-carboxylate
OR910533-1G OR910533-5G
APOSOR910533-1GEA 92.8 EUR
APOSOR910533-1G APOSOR910533-5G
Methyl-1-methyl-1H-1,3-benzimidazole-5-carboxylate 97%
Methyl-1-methyl-1H-1,3-benzimidazole-5-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04492113
CAS Number: 131020-36-5