N-Cyclopropyl-3-nitropyridin-2-amine 98%
Supplier: Apollo Scientific
Synonyms: N-cyclopropyl-3-nitropyridin-2-amine
OR911065-1G OR911065-5G
APOSOR911065-1GEA 111 EUR
APOSOR911065-1G APOSOR911065-5G
N-Cyclopropyl-3-nitropyridin-2-amine 98%
N-Cyclopropyl-3-nitropyridin-2-amine
Formula: C₈H₉N₃O₂
MW: 179.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174356
CAS Number: 290313-20-1