3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 98%
Supplier: Apollo Scientific
Synonyms: 1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)
OR911428-100G
APOSOR911428-100GEA 127 EUR
APOSOR911428-100G
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 98%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Formula: C₁₁H₁₈O₃
MW: 198.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006652
CAS Number: 69225-59-8