3-(3-Nitrophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR9115-1G OR9115-250MG
APOSOR9115-1GEA 46.4 EUR
APOSOR9115-1G APOSOR9115-250MG
3-(3-Nitrophenyl)-1H-pyrazole
3-(3-Nitrophenyl)-1H-pyrazole
Formula: C₉H₇N₃O₂
MW: 189.18 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00106146
CAS Number: 59843-77-5