2,3,5,6-Tetramethylpyrazine 98%
Supplier: Apollo Scientific
Warning
OR911661-500G
APOSOR911661-500GEA 122 EUR
APOSOR911661-500G
2,3,5,6-Tetramethylpyrazine 98%
2,3,5,6-Tetramethylpyrazine
Formula: C₈H₁₂N₂
MW: 136.2 g/mol
Boiling Pt: 190 °C/760mmHg
Melting Pt: 82…86 °C
Density: 1.08 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006146
CAS Number: 1124-11-4