Ethyl-2-cyano-4,4-diethoxybutyrate
Supplier: Apollo Scientific
Synonyms: Ethyl 2-cyano-4,4-diethoxybutyrate
OR911764-25G OR911764-5G
APOSOR911764-25GEA 108 EUR
APOSOR911764-25G APOSOR911764-5G
Ethyl-2-cyano-4,4-diethoxybutyrate
Ethyl-2-cyano-4,4-diethoxybutyrate
Formula: C₁₁H₁₉NO₄
MW: 229.2729 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094250
CAS Number: 52133-67-2
UN: 3276
ADR: 6.1,III