2,3,6,7-Tetrahydro-(1H)-1,4-diazepin-5-(4H)-one
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Diazepan-5-one, Homopiperazin-5-one
OR9118-1G OR9118-250MG OR9118-5G
APOSOR9118-1GEA 89.9 EUR
APOSOR9118-1G APOSOR9118-250MG APOSOR9118-5G
2,3,6,7-Tetrahydro-(1H)-1,4-diazepin-5-(4H)-one
2,3,6,7-Tetrahydro-(1H)-1,4-diazepin-5-(4H)-one
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00766804
CAS Number: 34376-54-0