3-Hydroxy-p-anisidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxy-4-anisidine, 5-Amino-2-methoxyphenol, 3-Hydroxy-4-methoxyaniline, 4-Amino-2-hydroxyanisole
OR911881-100G OR911881-25G
APOSOR911881-100GEA 282 EUR
APOSOR911881-100G APOSOR911881-25G
3-Hydroxy-p-anisidine 98%
3-Hydroxy-p-anisidine
Formula: C₇H₉NO₂
MW: 139.15 g/mol
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00010222
CAS Number: 1687-53-2