2-Chloro-3-(piperazin-1-yl)pyrazine 95%

Supplier: Apollo Scientific

OR911909-1G OR911909-250MG
APOSOR911909-1GEA 719 EUR
APOSOR911909-1G APOSOR911909-250MG
2-Chloro-3-(piperazin-1-yl)pyrazine 95%
2-Chloro-3-(piperazin-1-yl)pyrazine
Formula: C₈H₁₁ClN₄
Storage Temperature: Ambient
CAS Number: 85386-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR