1-Cyclopropylbutane-1,3-dione 97%
Supplier: Apollo Scientific
OR911927-100G OR911927-25G
APOSOR911927-100GEA 256 EUR
APOSOR911927-100G APOSOR911927-25G
1-Cyclopropylbutane-1,3-dione 97%
1-Cyclopropylbutane-1,3-dione
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00511094
CAS Number: 21573-10-4