1,1-Diphenylhydrazine hydrochloride 98%
Supplier: Apollo Scientific
Synonyms: N,N-Diphenylhydrazinium chloride
OR911932-25G OR911932-5G
APOSOR911932-25GEA 111 EUR
APOSOR911932-25G APOSOR911932-5G
1,1-Diphenylhydrazine hydrochloride 98%
1,1-Diphenylhydrazine hydrochloride
Formula: C₁₂H₁₃ClN₂
MW: 220.70 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012950
CAS Number: 530-47-2
UN: 3261
ADR: 8,III