L-Isoserine 97%

Supplier: Apollo Scientific

Synonyms: (S)-3-Amino-2-hydroxypropanoic acid, L-Isoserine, (S)-3-Amino-2-hydroxypropionic Acid

OR911994-100G
APOSOR911994-100GEA 121 EUR
APOSOR911994-100G
L-Isoserine 97%
L-Isoserine
Formula: C₃H₇NO₃
MW: 105.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03428326
CAS Number: 632-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR