DL-tert-Leucine (H-DL-Tle-OH) 97%
Supplier: Apollo Scientific
OR912027-100G OR912027-25G OR912027-5G
APOSOR912027-100GEA 219 EUR
APOSOR912027-100G APOSOR912027-25G APOSOR912027-5G
DL-tert-Leucine (H-DL-Tle-OH) 97%
DL-tert-Leucine (H-DL-Tle-OH)
Formula: C₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00065933
CAS Number: 33105-81-6