2,2-Dimethyl-3-hexanone 95%
Supplier: Apollo Scientific
Synonyms: 2,2-Dimethylhexan-3-one
OR912109-5G
APOSOR912109-5GEA 243 EUR
APOSOR912109-5G
2,2-Dimethyl-3-hexanone 95%
2,2-Dimethyl-3-hexanone
Formula: C₈H₁₆O
MW: 128.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00027140
CAS Number: 5405-79-8