3-Hydroxy-4-methylbenzonitrile 98%
Supplier: Apollo Scientific
OR912206-25G
APOSOR912206-25GEA 128 EUR
APOSOR912206-25G
3-Hydroxy-4-methylbenzonitrile 98%
3-Hydroxy-4-methylbenzonitrile
Formula: C₈H₇NO
MW: 133.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08361769
CAS Number: 3816-66-8