5'-Chloro-2'-methoxyacetophenone 95%
Supplier: Apollo Scientific
Synonyms: 1-(5-Chloro-2-methoxyphenyl)ethanone
OR912415-25G
APOSOR912415-25GEA 232 EUR
APOSOR912415-25G
5'-Chloro-2'-methoxyacetophenone 95%
5'-Chloro-2'-methoxyacetophenone
Formula: C₉H₉ClO₂
MW: 184.6196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174339
CAS Number: 6342-64-9
UN: 1759
ADR: 8,III