4-Amino-3-chlorophenol
Supplier: Apollo Scientific
Warning
OR912578-100G OR912578-25G OR912578-5G
APOSOR912578-100GEA 141 EUR
APOSOR912578-100G APOSOR912578-25G APOSOR912578-5G
4-Amino-3-chlorophenol
4-Amino-3-chlorophenol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00801116
CAS Number: 17609-80-2
UN: 2811
ADR: 6.1 III