3-Methylpyridine-4-carbaldehyde 95%
Supplier: Apollo Scientific
Danger
OR912752-1G OR912752-250MG
APOSOR912752-1GEA 470 EUR
APOSOR912752-1G APOSOR912752-250MG
3-Methylpyridine-4-carbaldehyde 95%
3-Methylpyridine-4-carbaldehyde
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 60…61 °C/0.5mmHg
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02181145
CAS Number: 74663-96-0