(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde 95%
Supplier: Apollo Scientific
Synonyms: 2,3-O-Cyclohexylidene-D-glyceraldehyde
OR912826-1G OR912826-5G
APOSOR912826-1GEA 144 EUR
APOSOR912826-1G APOSOR912826-5G
(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde 95%
(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde
Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093997
CAS Number: 78008-36-3